Novus Biologicals
Manufacturer Code:NBP2830100.1ML
Catalog # NB030939
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human EPS8L3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Epidermal growth factor receptor kinase substrate 8-like protein 3; epidermal growth factor receptor kinase substrate 8-like protein 3 Epidermal growth factor receptor pathway substrate 8-related protein 3 EPS8-like 3 EPS8-like protein 3 EPS8R3 EPS8-related protein 3 FLJ21522 MGC16817; Epidermal growth factor receptor pathway substrate 8-related protein 3; EPS8-like 3; EPS8-like protein 3; EPS8-related protein 3
Gene Aliases: EPS8L3; EPS8R3
UniProt ID: (Human) Q8TE67
Entrez Gene ID: (Human) 79574
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.