Proteintech
Manufacturer Code:204611AP150UL
Catalog # 50-173-2905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EPS8L2 Fusion Protein Ag14296 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Epidermal growth factor receptor kinase substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-like protein 2; Epidermal growth factor receptor pathway substrate 8-related protein 2; EPS8 like 2 EPS8 like protein 2 EPS8 related protein 2 EPS8L2 EPS8R2; EPS8 related protein 2; EPS8-like 2; EPS8-like protein 2; EPS8-related protein 2
Gene Aliases: AI042819; AW545405; EPS8L2; EPS8R2; PP13181
UniProt ID: (Human) Q9H6S3, (Mouse) Q99K30
Entrez Gene ID: (Human) 64787, (Mouse) 98845
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.