Novus Biologicals
Manufacturer Code:NBP15655720UL
Catalog # NBP15655720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to EPRS(glutamyl-prolyl-tRNA synthetase) The peptide sequence was selected from the middle region of EPRS. Peptide sequence KSEKQNKPQKQNDGQRKDPSKNQGGGLSSSGAGEGQGPKKQTRLGLEAKK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bifunctional aminoacyl-tRNA synthetase; bifunctional aminoacyl-tRNA synthetase Cell proliferation-inducing gene 32 protein DKFZp313B047 EARS EC 6.1.1 EC 6.1.1.15 EC 6.1.1.18 GLNS GLUPRORS glutamate tRNA ligase glutaminyl-tRNA synthetase glutamyl-prolyl-tRNA synthetase PARSproliferation-inducing gene 32 protein proliferation-inducing protein 32 proline tRNA ligase proline-tRNA ligase prolyl-tRNA synthetase QARS QPRS; Bifunctional glutamate/proline--tRNA ligase; Cell proliferation-inducing gene 32 protein; GluRS; glutamate tRNA ligase; Glutamate--tRNA ligase; Glutamatyl-prolyl-tRNA synthetase; glutaminyl-tRNA synthetase; Glutamyl-tRNA synthetase; proliferation-inducing gene 32 protein; proliferation-inducing protein 32; proline tRNA ligase; Proline--tRNA ligase; proline-tRNA ligase; Prolyl-tRNA synthetase
Gene Aliases: EARS; EPRS; EPRS1; GLNS; GLUPRORS; PARS; PIG32; QARS; QPRS
UniProt ID: (Human) P07814
Entrez Gene ID: (Human) 2058
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.