Abnova
Manufacturer Code:MAB14774
Catalog # 89-034-857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | EPO/1368 |
Immunogen | Recombinant protein corresponding to amino acids 28-162 of human EPO. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EP; Epoetin; Erythropoietin; MGC138142
Gene Aliases: EP; EPO; MVCD2
UniProt ID: (Human) P01588
Entrez Gene ID: (Human) 2056
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.