Abnova
Manufacturer Code:PAB18809
Catalog # 89-120-367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 890-904 of human EPHA4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of EPHA4.
For Research Use Only
Protein Aliases: EK8; EPH-like kinase 8; Ephrin type-A receptor 4; HEK8; receptor protein-tyrosine kinase HEK8; SEK; TYRO1; TYRO1 protein tyrosine kinase; Tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase TYRO1
Gene Aliases: EPHA4; HEK8; SEK; TYRO1
UniProt ID: (Human) P54764
Entrez Gene ID: (Human) 2043
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.