OriGene
Manufacturer Code:TA890117
Catalog # 50-168-4803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human EPHA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EK8; EPH-like kinase 8; Ephrin type-A receptor 4; HEK8 SEK TYRO1; receptor protein-tyrosine kinase HEK8; TYRO1 protein tyrosine kinase; Tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1
Gene Aliases: 2900005C20Rik; AI385584; Cek8; EPHA4; HEK8; rb; RGD1560587; SEK; Sek1; TYRO1
UniProt ID: (Human) P54764, (Mouse) Q80VZ2
Entrez Gene ID: (Human) 2043, (Rat) 316539, (Mouse) 13838
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.