Abnova
Manufacturer Code:H00002042M01
Catalog # 89-021-469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A12 |
Immunogen | EPHA3 (AAH63282 202 a.a. ~ 326 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant EPHA3.
For Research Use Only
Protein Aliases: EK4; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ETK; ETK1; HEK; HEK4; Human embryo kinase; human embryo kinase 1; testicular tissue protein Li 64; TYRO4; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase TYRO4
Gene Aliases: EK4; EPHA3; ETK; ETK1; HEK; HEK4; TYRO4
UniProt ID: (Human) P29320
Entrez Gene ID: (Human) 2042
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.