Invitrogen
Manufacturer Code:PA5115186
Catalog # PIPA5115186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EPHA3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW492086 Cek4 EK4 End3 Eph receptor A3 Epha3 EPH-like kinase 4 Eph-like tyrosine kinase 1 Ephrin type-A receptor 3 ETK ETK1 H TYRO4 HEK Hek4 Human embryo kinase human embryo kinase 1 Mek4 Rek4 testicular tissue protein Li 64 Tyro4 TYRO4 protein tyrosine kinase tyrosine-protein kinase receptor ETK1 tyrosine-protein kinase receptor REK4 tyrosine-protein kinase TYRO4; EK4; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; HEK; Human embryo kinase; human embryo kinase 1; testicular tissue protein Li 64; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase TYRO4
Gene Aliases: AW492086; Cek4; EK4; End3; EPHA3; ETK; ETK1; HEK; HEK4; Mek4; TYRO4
UniProt ID: (Human) P29320
Entrez Gene ID: (Human) 2042, (Mouse) 13837
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.