Abnova
Manufacturer Code:PAB27123
Catalog # 89-128-776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human EPB49. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of EPB49.
For Research Use Only
Protein Aliases: Dematin; Dematin actin-binding protein; DMT; Erythrocyte membrane protein band 4.9; erythrocyte membrane protein band 4.9 (dematin); FLJ78462; FLJ98848
Gene Aliases: DMT; DMTN; EPB49
UniProt ID: (Human) Q08495
Entrez Gene ID: (Human) 2039
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.