Abnova
Manufacturer Code:H00002039K
Catalog # 89-200-069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human EPB49 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human EPB49 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Dematin; Dematin actin-binding protein; DMT FLJ78462 FLJ98848; Erythrocyte membrane protein band 4.9; erythrocyte membrane protein band 4.9 (dematin)
Gene Aliases: DMT; DMTN; EPB49
UniProt ID: (Human) Q08495
Entrez Gene ID: (Human) 2039
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.