Novus Biologicals
Manufacturer Code:NBP287368
Catalog # NB034990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human EPB41L5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Band 4.1-like protein 5; band 4.1-like protein 5 erythrocyte membrane protein band 4.1 like 5 FLJ12957 KIAA1548BE37 YMO1; Erythrocyte membrane protein band 4.1-like 5; yurt homolog
Gene Aliases: BE37; EPB41L5; KIAA1548; YMO1; YRT
UniProt ID: (Human) Q9HCM4
Entrez Gene ID: (Human) 57669
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.