Proteintech
Manufacturer Code:107191AP150UL
Catalog # 50-172-6537
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EPB41L3 Fusion Protein Ag1089 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 130kDa Protein 4.1B MEF cell isoform; 4.1B; 4.1B Band 4.1 like protein 3 DAL 1 DAL1 EPB41L3 KIAA0987; 60kDa 4.1B MEF cell isoform; Band 4.1-like protein 3; Band 4.1-like protein 3, N-terminally processed; DAL-1; Differentially expressed in adenocarcinoma of the lung protein 1; Erythrocyte membrane protein band 4.1-like 3; erythrocyte protein band 4.1-like 3; mDAL-1
Gene Aliases: 4.1B; DAL-1; DAL1; DAL1P; Epb4.1l3; EPB41L3; KIAA0987; NBL3
UniProt ID: (Human) Q9Y2J2, (Mouse) Q9WV92
Entrez Gene ID: (Human) 23136, (Mouse) 13823, (Rat) 116724
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.