Invitrogen
Manufacturer Code:PA518830
Catalog # PIPA518830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DYDPEEHGSID) corresponding to the internal amino acids of EPB41L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 4.1G; Band 4.1-like protein 2; EPB41L2 erythrocyte membrane protein band 4.1-like 2 RP3-324N14.1 4.1-G DKFZp781D1972 DKFZp781H1755 OTTHUMP00000040262 erythrocyte membrane protein band 4.1 like-protein 2; erythrocyte membrane protein band 4.1 like-protein 2; Erythrocyte membrane protein band 4.1-like 2; Generally expressed protein 4.1
Gene Aliases: 4.1-G; 4.1G; EPB41L2
UniProt ID: (Human) O43491
Entrez Gene ID: (Human) 2037
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.