Proteintech
Manufacturer Code:154371AP150UL
Catalog # 50-173-0226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EPB41L2 Fusion Protein Ag3647 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4.1 G 4.1G Band 4.1 like protein 2 EPB41L2; 4.1G; Band 4.1-like protein 2; erythrocyte membrane protein band 4.1 like-protein 2; Erythrocyte membrane protein band 4.1-like 2; erythrocyte membrane protein band 4.1-like 2 retinal; erythrocyte protein band 4.1-like 2; erythrocyte protein band 4.1l2 long; erythrocyte protein band 4.1l2 middle; Generally expressed protein 4.1
Gene Aliases: 4.1-G; 4.1G; AW555191; D10Ertd398e; Epb4.1l2; EPB41L2; NBL2; RGD1563977
UniProt ID: (Human) O43491, (Mouse) O70318
Entrez Gene ID: (Human) 2037, (Mouse) 13822, (Rat) 309557
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.