Abnova
Manufacturer Code:H00002037K
Catalog # 89-204-989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human EPB41L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human EPB41L2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 4.1-G DKFZp781D1972 DKFZp781H1755; 4.1G; Band 4.1-like protein 2; erythrocyte membrane protein band 4.1 like-protein 2; Erythrocyte membrane protein band 4.1-like 2; Generally expressed protein 4.1
Gene Aliases: 4.1-G; 4.1G; EPB41L2
UniProt ID: (Human) O43491
Entrez Gene ID: (Human) 2037
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.