Proteintech
Manufacturer Code:191031AP150UL
Catalog # 50-173-2342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EPAC2 Fusion Protein Ag5186 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAMP GEFII CGEF2 EPAC 2 EPAC2 Nbla00496 RAPGEF4; cAMP-dependent Rap1 guanine-nucleotide exchange factor; cAMP-GEFII; cAMP-regulated guanine nucleotide exchange factor II; cAMP-regulated guanine nucleotide exchange factor IIB; EPAC 2; Exchange factor directly activated by cAMP 2; Exchange protein directly activated by cAMP 2; putative protein product of Nbla00496; Rap guanine nucleotide exchange factor (GEF) 4; Rap guanine nucleotide exchange factor 4; RAP guanine-nucleotide-exchange factor (GEF) 4
Gene Aliases: 1300003D15Rik; 5730402K07Rik; 6330581N18Rik; CAMP-GEFII; CAMPS; CGEF2; EPAC; EPAC 2; Epac-2; EPAC2; mKIAA4040; Nbla00496; RAPGEF4
UniProt ID: (Human) Q8WZA2, (Mouse) Q9EQZ6
Entrez Gene ID: (Human) 11069, (Rat) 252857, (Mouse) 56508
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.