Abnova
Manufacturer Code:MAB2385
Catalog # 89-110-980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | RW105 |
Immunogen | Recombinant protein corresponding to amino acids 1572-2371 of human EP300. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant EP300.
For Research Use Only
Protein Aliases: E1A-associated protein p300; E1A-binding protein, 300kD; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; KAT3B; p300; p300 HAT; Protein 2-hydroxyisobutyryltransferase p300; Protein lactyltransferas p300; Protein propionyltransferase p300
Gene Aliases: EP300; KAT3B; P300; RSTS2
UniProt ID: (Human) Q09472
Entrez Gene ID: (Human) 2033
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.