Invitrogen
Manufacturer Code:14487682
Catalog # 50-130-76
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 21Mags8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Eomesodermin homolog; Eomesodermin homolog EOMES TBR2; T-box brain protein 2; T-brain-2
Gene Aliases: C77258; EOMES; TBR-2; TBR2
UniProt ID: (Human) O95936, (Mouse) O54839
Entrez Gene ID: (Human) 8320, (Mouse) 13813
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.