Invitrogen
Manufacturer Code:PA534959
Catalog # PIPA534959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment contains a sequence corresponding to a region within amino acids 1 and 101 of ENY2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: enhancer of yellow 2 homolog; Enhancer of yellow 2 transcription factor homolog; Enhancer of yellow 2 transcription factor homolog DC6 ENY 2 ENY-2; ENY2; Transcription and mRNA export factor ENY2
Gene Aliases: DC6; e(y)2; ENY2; Sus1
UniProt ID: (Human) Q9NPA8
Entrez Gene ID: (Human) 56943
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.