Invitrogen
Manufacturer Code:PA5115182
Catalog # PIPA5115182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ENY2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810057B09Rik 6720481I12 DC6 e(y)2 e(y)2 homolog e(y)2 protein enhancer of yellow 2 homolog enhancer of yellow 2 homolog (Drosophila) enhancer of yellow 2 transcription factor homolog enhancer of yellow eye homolog ENY 2 ENY2 ENY-2 ENY2 transcription and export complex 2 subunit Ey2 Sus1 Transcription and mRNA export factor ENY2; e(y)2 homolog; e(y)2 protein; enhancer of yellow 2 homolog; Enhancer of yellow 2 transcription factor homolog; enhancer of yellow eye homolog; ENY2; Transcription and mRNA export factor ENY2
Gene Aliases: 1810057B09Rik; 6720481I12; DC6; e(y)2; ENY2; Ey2; Sus1
UniProt ID: (Human) Q9NPA8, (Mouse) Q9JIX0
Entrez Gene ID: (Human) 56943, (Rat) 685258, (Mouse) 223527
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.