Invitrogen
Manufacturer Code:PA5113799
Catalog # PIPA5113799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human ENTPD8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E-NTPDase 8; ecto-ATP-diphosphohydrolase Ectonucleoside triphosphate diphosphohydrolase 8 Entpd8 E-NTPDase E-NTPDase 8 GLSR2492 I79_018892 liver ecto-ATP-diphosphohydrolase NTPDase 8 NTPDase8 NTPDase-8 UNQ2492 UNQ2492/PRO5779; Ectonucleoside triphosphate diphosphohydrolase 8; liver ecto-ATP-diphosphohydrolase
Gene Aliases: E-NTPDase; ENTPD8; GLSR2492; NTPDase-8; UNQ2492; UNQ2492/PRO5779
UniProt ID: (Human) Q5MY95
Entrez Gene ID: (Human) 377841
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.