Novus Biologicals
Manufacturer Code:NBP159484
Catalog # NBP159484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ENTPD8(ectonucleoside triphosphate diphosphohydrolase 8) The peptide sequence was selected from the N terminal of ENTPD8. Peptide sequence IPEAQHRKTPTFLGATAGMRLLSRKNSSQARDIFAAVTQVLGRSPVDFWG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E-NTPDase 8; EC 3.6.1 EC 3.6.1.5 ecto-ATP-diphosphohydrolase ectonucleoside triphosphate diphosphohydrolase 8 E-NTPDase 8 GLSR2492 liver ecto-ATP-diphosphohydrolase NTPDase 8 NTPDase8 NTPDase-8 nucleoside triphosphate diphosphohydrolase-8 UNQ2492; Ectonucleoside triphosphate diphosphohydrolase 8; liver ecto-ATP-diphosphohydrolase
Gene Aliases: E-NTPDase; ENTPD8; GLSR2492; NTPDase-8; UNQ2492; UNQ2492/PRO5779
UniProt ID: (Human) Q5MY95
Entrez Gene ID: (Human) 377841
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.