Invitrogen
Manufacturer Code:PA597710
Catalog # PIPA597710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Ectonucleoside triphosphate diphosphohydrolase 7 protein (51-350AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ectonucleoside triphosphate diphosphohydrolase 7; lysosomal apyrase-like 2; Lysosomal apyrase-like protein 1; NTPDase 7
Gene Aliases: 1810012B13Rik; 1810020C02Rik; 2810003F23Rik; ENTPD7; Kiaa4066; LALP1; Lysal2
UniProt ID: (Human) Q9NQZ7, (Mouse) Q3TCT4
Entrez Gene ID: (Human) 57089, (Mouse) 93685
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.