Abnova
Manufacturer Code:H00000955M03
Catalog # 89-021-000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D10 |
Immunogen | ENTPD6 (NP_001238 386 a.a. ~ 484 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ENTPD6.
For Research Use Only
Protein Aliases: CD39 antigen-like 2; CD39L2; dJ738P15.3; DKFZp781G2277; DKFZp781K21102; Ectonucleoside triphosphate diphosphohydrolase 6; FLJ36711; IL-6SAG; IL6ST2; interleukin 6 signal transducer-2; NTPDase 6; NTPDase-6
Gene Aliases: CD39L2; dJ738P15.3; ENTPD6; IL-6SAG; IL6ST2; NTPDase-6
UniProt ID: (Human) O75354
Entrez Gene ID: (Human) 955
Molecular Function: hydrolase lyase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.