Invitrogen
Manufacturer Code:PA590599
Catalog # PIPA590599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 61-300 of human ENTPD6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 2; Ectonucleoside triphosphate diphosphohydrolase 6; NTPDase 6
Gene Aliases: 2700026H11Rik; Cd39l2; Entpd6; NTPDase-6
UniProt ID: (Mouse) Q3U0P5, (Rat) Q9ER31
Entrez Gene ID: (Mouse) 12497, (Rat) 85260
Molecular Function: hydrolase lyase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.