Invitrogen
Manufacturer Code:PA521393
Catalog # PIPA521393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 114 and 285 of ENTPD6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 2; Ectonucleoside triphosphate diphosphohydrolase 6; interleukin 6 signal transducer-2; NTPDase 6; RP4-738P15.5 CD39L2 IL-6SAG IL6ST2 NTPDase-6 dJ738P15.3 CD39 antigen-like 2 CD39-like 2 HCV-E2 binding protein 1 NTPDase 6 ectonucleoside triphosphate diphosphohydrolase 6 ectonucleoside triphosphate diphosphohydrolase 6 (putative function) interleukin 6 signal transducer-2
Gene Aliases: CD39L2; dJ738P15.3; ENTPD6; IL-6SAG; IL6ST2; NTPDase-6
UniProt ID: (Human) O75354
Entrez Gene ID: (Human) 955
Molecular Function:
hydrolase
lyase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.