Invitrogen
Manufacturer Code:MA529244
Catalog # PIMA529244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 19 |
Immunogen | Recombit Human ENTPD5 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 4; CD39-like 4; CD39L4 NTPDase-5 PCPH CD39 antigen-like 4 CD39-like 4 ER-UDPase GDPase ENTPD5 NTPDase 5 Pcph proto-oncogene protein UDPase ENTPD5 guanosine-diphosphatase ENTPD5 nucleoside diphosphatase proto-oncogene CPH uridine-diphosphatase ENTPD5; Ectonucleoside triphosphate diphosphohydrolase 5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; NTPDase 5; Nucleoside diphosphatase; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5
Gene Aliases: CD39L4; ENTPD5; NTPDase-5; PCPH
UniProt ID: (Human) O75356
Entrez Gene ID: (Human) 957
Molecular Function: hydrolase lyase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.