Invitrogen
Manufacturer Code:PA529503
Catalog # PIPA529503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 173 and 428 of Human ENTPD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 4; CD39L4 NTPDase-5 PCPH CD39 antigen-like 4 CD39-like 4 ER-UDPase GDPase ENTPD5 NTPDase 5 Pcph proto-oncogene protein UDPase ENTPD5 guanosine-diphosphatase ENTPD5 nucleoside diphosphatase proto-oncogene CPH uridine-diphosphatase ENTPD5; Ectonucleoside triphosphate diphosphohydrolase 5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; NTPDase 5; Nucleoside diphosphatase; UDPase ENTPD5; Uridine-diphosphatase ENTPD5
Gene Aliases: AI196558; AI987697; Cd39l4; Entpd5; ER-UDPase; mNTPase; NTPDase-5; NTPDase5; Pcph
UniProt ID: (Mouse) Q9WUZ9
Entrez Gene ID: (Mouse) 12499
Molecular Function: hydrolase lyase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.