Invitrogen
Manufacturer Code:PA5112412
Catalog # PIPA5112412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 5 protein (149-428AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI196558 AI987697 CD39 antigen-like 4 Cd39l4 CD39-like 4 Ectonucleoside triphosphate diphosphohydrolase 5 ENTPD5 ER-UDPase GDPase ENTPD5 Guanosine-diphosphatase ENTPD5 mNTPase NTPDase 5 NTPDase5 NTPDase-5 Nucleoside diphosphatase PCPH Pcph proto-oncogene protein proto-oncogene CPH UDPase ENTPD5 Uridine-diphosphatase ENTPD5; CD39 antigen-like 4; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; NTPDase 5; Nucleoside diphosphatase; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5
Gene Aliases: AI196558; AI987697; CD39L4; ENTPD5; ER-UDPase; mNTPase; NTPDase-5; NTPDase5; PCPH
UniProt ID: (Human) O75356, (Mouse) Q9WUZ9
Entrez Gene ID: (Human) 957, (Mouse) 12499
Molecular Function:
hydrolase
lyase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.