Novus Biologicals
Manufacturer Code:NBP181510
Catalog # NBP181510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:FGGNAARQRYEDRIFANTIQKNRLLGKQTGLTPDMPYLDPCLPLDIKDEIQQNGQTIYLRGTGDFDLCRETIQPFMNKTNETQTSLNGVYQPPIHFQNSEFYGFSEFYYCTEDVLR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ectonucleoside triphosphate diphosphohydrolase 4; ectonucleoside triphosphate diphosphohydrolase 4 KIAA0392NTPDase 4 LALP70EC 3.6.1.6 LAP70guanosine-diphosphatase like protein LYSAL1uridine-diphosphatase lysosomal apyrase-like 1 Lysosomal apyrase-like protein of 70 kDa NTPDase-4 UDPaseapyrase lysosomal Uridine-diphosphatase; golgi luminal UDPase; Golgi UDPase; guanosine-diphosphatase like protein; lysosomal apyrase-like 1; Lysosomal apyrase-like protein of 70 kDa; NTPDase 4; UDPase; Uridine-diphosphatase
Gene Aliases: ENTPD4; KIAA0392; LALP70; LAP70; LYSAL1; NTPDase-4; UDPase
UniProt ID: (Human) Q9Y227
Entrez Gene ID: (Human) 9583
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.