Proteintech
Manufacturer Code:195971AP150UL
Catalog # 50-173-2463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ENTPD4 Fusion Protein Ag5175 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ectonucleoside triphosphate diphosphohydrolase 4; ENTPD4 KIAA0392 LALP70 LAP70 LYSAL1 NTPDase 4 UDPase Uridine diphosphatase; golgi luminal UDPase; Golgi UDPase; guanosine-diphosphatase like protein; lysosomal apyrase-like 1; Lysosomal apyrase-like protein of 70 kDa; NTPDase 4; UDPase; Uridine-diphosphatase
Gene Aliases: 1200014F22Rik; 4930513O14Rik; A430102M06Rik; AI414893; ENTPD4; KIAA0392; LALP70; LAP70; LYSAL1; mKIAA0392; NTPDase-4; UDPase
UniProt ID: (Human) Q9Y227, (Mouse) Q9DBT4
Entrez Gene ID: (Human) 9583, (Rat) 361063, (Mouse) 67464
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.