Proteintech
Manufacturer Code:130211AP150UL
Catalog # 50-172-8424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ENTPD3 Fusion Protein Ag4114 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen like 3 CD39L3 Ecto apyrase 3 Ecto ATP diphosphohydrolase 3 Ecto ATPase 3 Ecto ATPDase 3 ENTPD3 HB6 NTPDase 3; CD39 antigen-like 3; Ecto-apyrase 3; Ecto-ATP diphosphohydrolase 3; ecto-ATPase 3; Ecto-ATPDase 3; Ectonucleoside triphosphate diphosphohydrolase 3; HB6; NTPDase 3
Gene Aliases: CD39L3; ENTPD3; HB6; NTPDase-3; NTPDase3
UniProt ID: (Human) O75355
Entrez Gene ID: (Human) 956, (Rat) 316077, (Mouse) 215446
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.