Invitrogen
Manufacturer Code:PA532751
Catalog # PIPA532751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from internal region of human ENTPD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 1; CD39L1 NTPDase-2 CD39 antigen-like 1 NTPDase 2 ecto-ATP diphosphohydrolase 2 ecto-ATPDase 2 ecto-ATPase 2; Ecto-ATP diphosphohydrolase 2; ecto-ATPase 2; Ecto-ATPDase 2; Ectonucleoside triphosphate diphosphohydrolase 2; NTPDase 2
Gene Aliases: CD39L1; ENTPD2; NTPDase-2
UniProt ID: (Human) Q9Y5L3
Entrez Gene ID: (Human) 954
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.