Invitrogen
Manufacturer Code:PA577545
Catalog # PIPA577545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EPQKPGKPSVFVVFRK corresponding to amino acid residues 273-288 of rat Equilibrative Nucleoside Transporter 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 36 kDa hydrophobic nucleolar protein; 36 kDa nucleolar protein HNP36; delayed early response gene 12; Delayed-early response protein 12; Equilibrative NBMPR-insensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleoside transporter; Equilibrative nucleoside transporter 2; Nucleoside transporter ei-type SLC29A2; Nucleoside transporter, ei-type; solute carrier family 29 (nucleoside transporters), member 2; Solute carrier family 29 member 2; solute carrier family 29, member 2
Gene Aliases: Der12; Ent2; Hnp36; Slc29a2
UniProt ID: (Mouse) Q61672, (Rat) O54699
Entrez Gene ID: (Mouse) 13340, (Rat) 65194
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.