Invitrogen
Manufacturer Code:PA553746
Catalog # PIPA553746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC29A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 36 kDa nucleolar protein HNP36; Delayed-early response protein 12; Equilibrative NBMPR-insensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleoside transporter; Equilibrative nucleoside transporter 2; Hydrophobic nucleolar protein, 36 kDa; hydrophobic nucleolar protein, 36kD; Nucleoside transporter, ei-type; solute carrier family 29 (equilibrative nucleoside transporter), member 2; solute carrier family 29 (nucleoside transporters), member 2; Solute carrier family 29 member 2; Solute carrier family 29 member 2 Equilibrative nucleoside transporter 2 ENT2 DER12 HNP36
Gene Aliases: DER12; ENT2; HNP36; SLC29A2
UniProt ID: (Human) Q14542
Entrez Gene ID: (Human) 3177
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.