Invitrogen
Manufacturer Code:PA575507
Catalog # PIPA575507
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 5-50 of Human ENT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Equilibrative NBMPR-sensitive nucleoside transporter; equilibrative nitrobenzylmercaptopurine riboside (NBMPR)-sensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter; Equilibrative nucleoside transporter 1; Equilibrative nucleoside transporter 1 Equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter Equilibrative NBMPR-sensitive nucleoside transporter Nucleoside transporter es-type Solute carrier family 29 member 1 ENT1; equilibrative nucleoside transporter 1 variant delta 11; NBMPR-sensitive equilibrative nucleoside transporter; Nucleoside transporter, es-type; solute carrier family 29 (equilibrative nucleoside transporter), member 1; solute carrier family 29 (nucleoside transporters), member 1; Solute carrier family 29 member 1; solute carrier family 29, member 1
Gene Aliases: 1200014D21Rik; AA407560; ENT1; rENT1; SLC29A1
UniProt ID: (Human) Q99808, (Mouse) Q9JIM1, (Rat) O54698
Entrez Gene ID: (Human) 2030, (Mouse) 63959, (Rat) 63997
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.