Invitrogen
Manufacturer Code:PA5111872
Catalog # PIPA5111872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 245-260 of human ENT1 (3rdintracellular loop) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200014D21Rik AA407560 Ent1 Equilibrative NBMPR-sensitive nucleoside transporter Equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter Equilibrative nucleoside transporter 1 HENT1 Nucleoside transporter es-type rENT1 SLC29A1 Solute carrier family 29 member 1; Equilibrative NBMPR-sensitive nucleoside transporter; equilibrative nitrobenzylmercaptopurine riboside (NBMPR)-sensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter; Equilibrative nucleoside transporter 1; NBMPR-sensitive equilibrative nucleoside transporter; Nucleoside transporter, es-type; solute carrier family 29 (equilibrative nucleoside transporter), member 1; solute carrier family 29 (nucleoside transporters), member 1; Solute carrier family 29 member 1; solute carrier family 29, member 1
Gene Aliases: ENT1; rENT1; SLC29A1
UniProt ID: (Human) Q99808, (Rat) O54698
Entrez Gene ID: (Human) 2030, (Rat) 63997
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.