Proteintech
Manufacturer Code:145181AP150UL
Catalog # 50-172-9508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ENSA Fusion Protein Ag5982 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha endosulfine ARPP 19e endosulfine alpha ENSA; Alpha-endosulfine; ARPP-19e; endosulfine-alpha variant 1; endosulfine-alpha variant 3
Gene Aliases: 1700020C18Rik; 2610007F17Rik; AI451924; ARPP-19e; ENSA
UniProt ID: (Human) O43768, (Rat) P60841, (Mouse) P60840
Entrez Gene ID: (Human) 2029, (Rat) 60334, (Mouse) 56205
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.