Proteintech
Manufacturer Code:126431AP150UL
Catalog # 50-172-8139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ENPP6 Fusion Protein Ag3339 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B830047L21Rik; Choline-specific glycerophosphodiester phosphodiesterase; E NPP 6 ENPP6; E-NPP 6; E-NPP6; Ectonucleotide pyrophosphatase/phosphodiesterase family member 6; glycerophosphocholine cholinephosphodiesterase; Glycerophosphocholine cholinephosphodiesterase ENPP6; GPC-Cpde; NPP-6
Gene Aliases: 4833421B01Rik; B830047L21Rik; D8Ertd514e; E-NPP 6; ENPP6; NPP6; UNQ1889/PRO4334
UniProt ID: (Human) Q6UWR7, (Mouse) Q8BGN3, (Rat) B0BND0
Entrez Gene ID: (Human) 133121, (Mouse) 320981, (Rat) 306460
Molecular Function: hydrolase nucleotide phosphatase phosphatase pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.