Novus Biologicals
Manufacturer Code:NBP305178100UL
Catalog # NB095276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 90-230 of human ENPP4 (NP_055751.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AP3A hydrolase; AP3Aase; Bis(5'-adenosyl)-triphosphatase ENPP4; E-NPP 4; EC 3.1 ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative) E-NPP 4 KIAA0879ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative function) NPP-4 NPP4ectonucleotide pyrophosphatase/phosphodiesterase family member 4; ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative function); Ectonucleotide pyrophosphatase/phosphodiesterase family member 4; NPP-4
Gene Aliases: ENPP4; KIAA0879; NPP4
UniProt ID: (Human) Q9Y6X5
Entrez Gene ID: (Human) 22875
Molecular Function: hydrolase nucleotide phosphatase phosphatase pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.