Proteintech
Manufacturer Code:173051AP150UL
Catalog # 50-173-1499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ENPP4 Fusion Protein Ag11192 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AP3A hydrolase; AP3Aase; Bis(5'-adenosyl)-triphosphatase ENPP4; E NPP 4 ENPP4 KIAA0879; E-NPP 4; E-NPP4; ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative function); Ectonucleotide pyrophosphatase/phosphodiesterase family member 4; NPP-4
Gene Aliases: 4933413N07Rik; AA986363; AI195364; E-NPP 4; ENPP4; Gm90; KIAA0879; mKIAA0879; NPP4
UniProt ID: (Human) Q9Y6X5, (Mouse) Q8BTJ4
Entrez Gene ID: (Human) 22875, (Rat) 301261, (Mouse) 224794
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.