Abnova
Manufacturer Code:H00022875K
Catalog # 89-206-769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ENPP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ENPP4 peptide using ARM Technology.
For Research Use Only
Protein Aliases: AP3A hydrolase; AP3Aase; Bis(5'-adenosyl)-triphosphatase ENPP4; E-NPP 4; ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative function); Ectonucleotide pyrophosphatase/phosphodiesterase family member 4; KIAA0879 NPP4; NPP-4
Gene Aliases: ENPP4; KIAA0879; NPP4
UniProt ID: (Human) Q9Y6X5
Entrez Gene ID: (Human) 22875
Molecular Function: hydrolase nucleotide phosphatase phosphatase pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.