Invitrogen
Manufacturer Code:PA545041
Catalog # PIPA545041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ENOX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APK1 antigen; Cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; Ecto-NOX disulfide-thiol exchanger 2 APK1 COVA1 tNOX; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; tNOX; Tumor-associated hydroquinone oxidase; zmp:0000001197
Gene Aliases: APK1; COVA1; ENOX2; RGD1563643; tNOX
UniProt ID: (Human) Q16206, (Mouse) Q8R0Z2
Entrez Gene ID: (Dog) 481050, (Horse) 100058391, (Human) 10495, (Rabbit) 100348937, (Mouse) 209224, (Rat) 302817, (Zebrafish) 566327
Molecular Function:
nuclease
nucleic acid binding
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.