Abnova
Manufacturer Code:H00010495A01
Catalog # 89-106-333
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 309 a.a. to 379 a.a) partial recombinant protein with GST tag.,ENOX2 (NP_872114 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ENOX2.
For Research Use Only
Protein Aliases: APK1; APK1 antigen; COVA1; Cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; tNOX; Tumor-associated hydroquinone oxidase
Gene Aliases: APK1; COVA1; ENOX2; tNOX
UniProt ID: (Human) Q16206
Entrez Gene ID: (Human) 10495
Molecular Function:
nuclease
nucleic acid binding
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.