Invitrogen
Manufacturer Code:PA560776
Catalog # PIPA560776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ENOPH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2,3-diketo-5-methylthio-1-phosphopentane phosphatase; acireductone synthase; E1 MASA MST145 23-diketo-5-methylthio-1-phosphopentane phosphatase MASA homolog acireductone synthase enolase-phosphatase E1; Enolase-phosphatase E1; ENOPH1; MASA homolog
Gene Aliases: E1; ENOPH1; MASA; MST145; MSTP145; mtnC
UniProt ID: (Human) Q9UHY7
Entrez Gene ID: (Human) 58478
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.