Invitrogen
Manufacturer Code:PA5100501
Catalog # PIPA5100501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ENOPH1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2,3-diketo-5-methylthio-1-phosphopentane phosphatase; 2,3-diketo-5-methylthio-1-phosphopentane phosphatase {ECO:0000255|HAMAP-Rule:MF_03117}; 23-diketo-5-methylthio-1-phosphopentane phosphatase 23-diketo-5-methylthio-1-phosphopentane phosphatase {ECO:0000255 2310057D15Rik acireductone synthase BB183658 C81437 E1 E-1 enzyme enolase-phosphatase 1 enolase-phosphatase E1 enolase-phosphatase E1 {ECO:0000255 Enoph1 HAMAP-Rule:MF_03117} MASA MASA homolog MASA homolog {ECO:0000255 MST145 MSTP145 mtnC RGD1309016; acireductone synthase; E-1 enzyme; Enolase-phosphatase E1; enolase-phosphatase E1 {ECO:0000255|HAMAP-Rule:MF_03117}; ENOPH1; MASA homolog; MASA homolog {ECO:0000255|HAMAP-Rule:MF_03117}
Gene Aliases: 2310057D15Rik; BB183658; C81437; E1; ENOPH1; MASA; MST145; MSTP145; mtnC; RGD1309016
UniProt ID: (Human) Q9UHY7, (Mouse) Q8BGB7, (Rat) Q5PPH0
Entrez Gene ID: (Human) 58478, (Mouse) 67870, (Rat) 305177
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.