OriGene
Manufacturer Code:CF506170
Catalog # 50-166-4208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8A1 |
Immunogen | Full length human recombit protein of human ENG produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD105; CD105 antigen; Cell surface MJ7/18 antigen; END HHT1 ORW1; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); transmembrane glycoprotein
Gene Aliases: AI528660; AI662476; CD105; Edg; END; Endo; ENG; HHT1; ORW1; S-endoglin
UniProt ID: (Human) P17813, (Mouse) Q63961
Entrez Gene ID: (Human) 2022, (Dog) 609166, (Mouse) 13805, (Rat) 497010
Molecular Function: TGF-beta receptor cytokine receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.