Invitrogen
Manufacturer Code:PA530018
Catalog # PIPA530018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 30 and 264 of Human Endo G |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endo G; endo G endonuclease G mitochondrial mitochondrial endonuclease G; Endonuclease G, mitochondrial; mitochondrial endonuclease G
Gene Aliases: ENDOG
UniProt ID: (Human) Q14249, (Mouse) O08600
Entrez Gene ID: (Human) 2021, (Mouse) 13804
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.