Proteintech
Manufacturer Code:550321AP150UL
Catalog # 50-173-5946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: empty spiracles homeobox 1; empty spiracles homeobox 1 Empty spiracles homolog 1 Empty spiracles like protein 1 EMX1 Homeobox protein EMX1; Empty spiracles homolog 1; Empty spiracles-like protein 1; Homeobox protein EMX1
Gene Aliases: Emx-1; EMX1; RGD1564002
UniProt ID: (Human) Q04741, (Mouse) Q04742
Entrez Gene ID: (Human) 2016, (Rat) 500235, (Mouse) 13796
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.