Invitrogen
Manufacturer Code:PA547914
Catalog # PIPA547914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human EMP/MAEA Met12-Ser66 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 5 protein; E3 ubiquitin-protein transferase MAEA; EMLP EMP HLC-10 PIG5 cell proliferation-inducing gene 5 protein erythroblast macrophage protein human lung cancer oncogene 10 protein lung cancer-related protein 10 proliferation-inducing gene 5; Erythroblast macrophage protein; GID complex subunit 9, FYV10 homolog; HLC-10; Human lung cancer oncogene 10 protein; lung cancer-related protein 10; Macrophage erythroblast attacher; P44EMLP
Gene Aliases: 1110030D19Rik; EMLP; EMP; GID9; HLC-10; HLC10; MAEA; PIG5
UniProt ID: (Human) Q7L5Y9, (Rat) Q5RKJ1, (Mouse) Q4VC33
Entrez Gene ID: (Human) 10296, (Rat) 298982, (Mouse) 59003
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.